6-Bromo-1-benzothiophene-2-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Bromobenzo[b]thiophene-2-carboxylic acid

26417-25G 26417-5G
76938-870EA 630.18 USD
76938-870 76938-872
6-Bromo-1-benzothiophene-2-carboxylic acid ≥97%
6-Bromo-1-benzothiophene-2-carboxylic acid
Formula: C₉H₅BrO₂S
MW: 257.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07371540
CAS Number: 19075-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR