5-Methoxy-2-methylphenol ≥95%
Supplier: AOBChem USA
|
Warning
|
26402-25G
26402-5G
76938-862EA
399.96
USD
76938-862
76938-864
5-Methoxy-2-methylphenol ≥95%
5-Methoxy-2-methylphenol
Precaución: for Research Use Only
Formula:
C₈H₁₀O₂ Storage Temperature: Ambient |
MDL Number:
MFCD12405727 CAS Number: 20734-74-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...