2-Fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Fluoropyridine-2-boronic acid pinacol ester

26302-1G 26302-25G 26302-500MG 26302-5G
76938-836EA 281.14 USD
76938-836 76938-838 76938-840 76938-842
2-Fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine ≥97%
2-Fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Formula: C₁₁H₁₅BFNO₂
MW: 223.05 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08063029
CAS Number: 842136-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR