N-(3-Bromobenzyl)-N-(tert-butyl)amine ≥97%
Supplier: AOBChem USA
|
Warning
|
26200-1G
26200-25G
26200-500MG
26200-5G
76938-822EA
138.98
USD
76938-822
76938-824
76938-826
76938-828
N-(3-Bromobenzyl)-N-(tert-butyl)amine ≥97%
N-(3-Bromobenzyl)-N-(tert-butyl)amine
Precaución: for Research Use Only
Formula:
C₁₁H₁₆BrN MW: 242.155 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD04526045 CAS Number: 133042-85-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...