Methyl 2-fluoro-5-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

26131-1G 26131-25G 26131-5G
76938-808EA 64.72 USD
76938-808 76938-810 76938-812
Methyl 2-fluoro-5-methoxybenzoate ≥97%
Methyl 2-fluoro-5-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₉FO₃
MW: 184.164 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06203711
CAS Number: 96826-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR