7-Methoxy-1H-indazole-4-boronic acid ≥97%

Supplier: AOBChem USA

Warning

26117-1G 26117-500MG 26117-5G
76938-794EA 485.9 USD
76938-794 76938-796 76938-798
7-Methoxy-1H-indazole-4-boronic acid ≥97%
7-Methoxy-1H-indazole-4-boronic acid

Precaución: for Research Use Only

Formula: C₈H₉BN₂O₃
MW: 191.98 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26045255
CAS Number: 2121513-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR