Methyl 2-chloro-4-methylnicotinate ≥97%

Supplier: AOBChem USA

Warning

26106-1G 26106-25G 26106-500MG 26106-5G
76938-778EA 411.63 USD
76938-778 76938-780 76938-782 76938-784
Methyl 2-chloro-4-methylnicotinate ≥97%
Methyl 2-chloro-4-methylnicotinate

Precaución: for Research Use Only

Formula: C₈H₈ClNO₂
MW: 185.608 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD15146644
CAS Number: 217811-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR