5-Methyl-2-phenylaniline ≥97%
Supplier: AOBChem USA
|
Warning
|
26029-1G
26029-25G
26029-500MG
26029-5G
76938-764EA
168.69
USD
76938-764
76938-766
76938-768
76938-770
5-Methyl-2-phenylaniline ≥97%
5-Methyl-2-phenylaniline
Precaución: for Research Use Only
Formula:
C₁₃H₁₃N Storage Temperature: Refrigerator |
MDL Number:
MFCD18970945 CAS Number: 54147-94-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...