6-Hydrazinyl-1H-pyrazolo[3,4-b]pyridine ≥97%

Supplier: AOBChem USA

Warning

26012-1G 26012-500MG 26012-5G
76938-754EA 528.33 USD
76938-754 76938-756 76938-758
6-Hydrazinyl-1H-pyrazolo[3,4-b]pyridine ≥97%
6-Hydrazinyl-1H-pyrazolo[3,4-b]pyridine

Precaución: for Research Use Only

Formula: C₆H₇N₅
MW: 149.153 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13178165
CAS Number: 918485-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR