6-Bromo-2-fluoro-3-iodophenol ≥97%

Supplier: AOBChem USA

Warning

25948-1G 25948-25G 25948-5G
76938-730EA 84.88 USD
76938-730 76938-732 76938-734
6-Bromo-2-fluoro-3-iodophenol ≥97%
6-Bromo-2-fluoro-3-iodophenol

Precaución: for Research Use Only

Formula: C₆H₃BrFIO
Storage Temperature: Ambient
MDL Number: MFCD24848713
CAS Number: 1449008-11-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR