2-Chloro-4-iodonicotinic acid methyl ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 2-chloro-4-iodonicotinate, Methyl 2-chloro-4-iodopyridine-3-carboxylate

25919-1G 25919-25G 25919-5G
76938-706EA 71.09 USD
76938-706 76938-708 76938-710
2-Chloro-4-iodonicotinic acid methyl ester ≥97%
2-Chloro-4-iodonicotinic acid methyl ester
Formula: C₇H₅ClINO₂
MW: 297.48 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04039871
CAS Number: 185041-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR