6-Bromo-2-fluoro-3-iodobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

25863-1G 25863-25G 25863-5G
76938-692EA 71.09 USD
76938-692 76938-694 76938-696
6-Bromo-2-fluoro-3-iodobenzaldehyde ≥97%
6-Bromo-2-fluoro-3-iodobenzaldehyde
Formula: C₇H₃BrFIO
Storage Temperature: Ambient
MDL Number: MFCD23713164
CAS Number: 1428234-59-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR