5-Phenylnicotinaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 5-Phenylpyridine-3-carboxaldehyde

25781-1G 25781-25G 25781-500MG 25781-5G
76938-670EA 228.1 USD
76938-670 76938-672 76938-674 76938-676
5-Phenylnicotinaldehyde ≥97% research grade
5-Phenylnicotinaldehyde
Formula: C₁₂H₉NO
MW: 183.21 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03086178
CAS Number: 113118-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR