7-Iodo-1H-indazole ≥97%
Supplier: AOBChem USA
|
Warning
|
25725-1G
25725-25G
25725-500MG
25725-5G
76938-654EA
126.25
USD
76938-654
76938-656
76938-658
76938-660
7-Iodo-1H-indazole ≥97%
7-Iodo-1H-indazole
Formula:
C₇H₅IN₂ Storage Temperature: Ambient |
MDL Number:
MFCD09263215 CAS Number: 945761-94-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...