Methyl 4-bromo-2-chloronicotinate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 4-bromo-2-chloropyridine-3-carboxylate

25705-1G 25705-25G 25705-5G
76938-644EA 73.21 USD
76938-644 76938-646 76938-648
Methyl 4-bromo-2-chloronicotinate ≥97%
Methyl 4-bromo-2-chloronicotinate
Formula: C₇H₅BrClNO₂
MW: 250.47 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD21364445
CAS Number: 1064678-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR