5-Methyl-1H-pyrazolo[3,4-b]pyridine ≥97%
Supplier: AOBChem USA
|
Warning
|
25541-1G
25541-25G
25541-500MG
25541-5G
76938-620EA
279.02
USD
76938-620
76938-622
76938-624
76938-626
5-Methyl-1H-pyrazolo[3,4-b]pyridine ≥97%
5-Methyl-1H-pyrazolo[3,4-b]pyridine
Precaución: for Research Use Only
Formula:
C₇H₇N₃ MW: 133.151 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD17676407 CAS Number: 1426427-07-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...