7-Chloro-4-fluoro-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

25425-1G 25425-25G 25425-500MG 25425-5G
76938-608EA 266.29 USD
76938-608 76938-610 76938-612 76938-614
7-Chloro-4-fluoro-1H-indazole ≥97%
7-Chloro-4-fluoro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄ClFN₂
MW: 170.571 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09749913
CAS Number: 1000341-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR