tert-Butyl 3-bromo-5-fluorobenzoate ≥95% research grade

Supplier: AOBChem USA

Warning

25397-1G 25397-25G 25397-500MG 25397-5G
76938-592EA 198.39 USD
76938-592 76938-594 76938-596 76938-598
tert-Butyl 3-bromo-5-fluorobenzoate ≥95% research grade
tert-Butyl 3-bromo-5-fluorobenzoate
Formula: C₁₁H₁₂BrFO₂
MW: 275.11 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD19388677
CAS Number: 1262834-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR