2-Phenyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4,4,5,5-Tetramethyl-2-(6-phenyl-3-pyridyl)-1,3,2-dioxaborolane, 6-Phenylpyridine-3-boronic acid pinacol ester, 6-Phenyl-3-pyridylboronic acid Pinacol ester

25282-1G 25282-25G 25282-500MG 25282-5G
76938-574EA 173.99 USD
76938-574 76938-576 76938-578 76938-580
2-Phenyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine ≥97%
2-Phenyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₁₇H₂₀BNO₂
Storage Temperature: Ambient
MDL Number: MFCD12032565
CAS Number: 879291-27-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR