Methyl 3-fluoro-5-methoxybenzoate ≥95%

Supplier: AOBChem USA

Warning

25103-1G 25103-25G 25103-500MG 25103-5G
76938-558EA 195.21 USD
76938-558 76938-560 76938-562 76938-564
Methyl 3-fluoro-5-methoxybenzoate ≥95%
Methyl 3-fluoro-5-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₉FO₃
MW: 184.164 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698049
CAS Number: 1214387-14-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR