6-Chloro-3-iodo-1H-indazole ≥95%
Supplier: AOBChem USA
|
Warning
|
25016-1G
25016-25G
25016-500MG
25016-5G
76938-548EA
173.99
USD
76938-548
76938-550
76938-552
76938-554
6-Chloro-3-iodo-1H-indazole ≥95%
6-Chloro-3-iodo-1H-indazole
Precaución: for Research Use Only
Formula:
C₇H₄ClIN₂ MW: 278.478 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD07781533 CAS Number: 503045-59-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...