6-Isobutoxy-2-methylpyridine-3-boronic acid ≥97%

Supplier: AOBChem USA

Warning

24835-1G 24835-25G 24835-500MG 24835-5G
76938-520EA 170.81 USD
76938-520 76938-522 76938-524 76938-526
6-Isobutoxy-2-methylpyridine-3-boronic acid ≥97%
6-Isobutoxy-2-methylpyridine-3-boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₆BNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD21332944
CAS Number: 1451390-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR