6-Isopropoxynaphthalene-2-boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

24751-1G 24751-25G 24751-5G
76938-508EA 78.51 USD
76938-508 76938-510 76938-512
6-Isopropoxynaphthalene-2-boronic acid ≥97% research grade
6-Isopropoxynaphthalene-2-boronic acid
Formula: C₁₃H₁₅BO₃
Storage Temperature: Refrigerator
MDL Number: MFCD16260719
CAS Number: 1334221-34-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR