6-Fluoro-3-iodo-2-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

24486-1G 24486-25G 24486-500MG 24486-5G
76938-494EA 409.51 USD
76938-494 76938-496 76938-498 76938-500
6-Fluoro-3-iodo-2-methoxybenzaldehyde ≥97%
6-Fluoro-3-iodo-2-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆FIO₂
MW: 280.035 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22543708
CAS Number: 1394291-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR