6-Phenoxy-3-formylpyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Phenoxynicotinaldehyde, 6-Phenoxypyridine-3-carboxaldehyde

24388-1G 24388-25G 24388-500MG 24388-5G
76938-480EA 135.8 USD
76938-480 76938-482 76938-484 76938-486
6-Phenoxy-3-formylpyridine ≥97%
6-Phenoxy-3-formylpyridine

Precaución: for Research Use Only

Formula: C₁₂H₉NO₂
MW: 199.21 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02681950
CAS Number: 173282-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR