Methyl 5-bromo-4-methylnicotinate ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: Methyl 5-Bromo-4-methylpyridine-3-carboxylate

24276-1G 24276-25G 24276-500MG 24276-5G
76938-464EA 260.99 USD
76938-464 76938-466 76938-468 76938-470
Methyl 5-bromo-4-methylnicotinate ≥97% research grade
Methyl 5-bromo-4-methylnicotinate
Formula: C₈H₈BrNO₂
MW: 230.06 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22543688
CAS Number: 1382847-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR