Methyl 3,5-dibromo-2-carboxylate ≥95%

Supplier: AOBChem USA

Warning

24205-1G 24205-25G 24205-500MG 24205-5G
76938-450EA 260.99 USD
76938-450 76938-452 76938-454 76938-456
Methyl 3,5-dibromo-2-carboxylate ≥95%
Methyl 3,5-dibromo-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₄Br₂O₂S
MW: 299.968 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22380015
CAS Number: 62224-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR