Methyl 2-chloro-6-hydroxybenzoate ≥97%

Supplier: AOBChem USA

Warning

24193-1G 24193-25G 24193-500MG 24193-5G
76938-434EA 201.58 USD
76938-434 76938-436 76938-438 76938-440
Methyl 2-chloro-6-hydroxybenzoate ≥97%
Methyl 2-chloro-6-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₈H₇ClO₃
MW: 186.592 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15523524
CAS Number: 176750-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR