Quinolin-3-ylmethanol ≥97%

Supplier: AOBChem USA

Warning

24121-1G 24121-25G 24121-500MG 24121-5G
76938-418EA 129.43 USD
76938-418 76938-420 76938-422 76938-424
Quinolin-3-ylmethanol ≥97%
Quinolin-3-ylmethanol
Formula: C₁₀H₉NO
Storage Temperature: Refrigerator
MDL Number: MFCD03412552
CAS Number: 13669-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR