N-Benzyl-5-bromonicotinamide ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: N-Benzyl-5-bromonicatinamide

24076-25G 24076-5G
76938-394EA 321.46 USD
76938-394 76938-396
N-Benzyl-5-bromonicotinamide ≥97% research grade
N-Benzyl-5-bromonicotinamide
Formula: C₁₃H₁₁BrN₂O
MW: 291.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01788380
CAS Number: 303031-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR