6-Fluoro-4-iodo-3-pyridinol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Fluoro-4-iodopyridin-3-ol

24029-1G 24029-25G 24029-500MG 24029-5G
76938-376EA 336.31 USD
76938-376 76938-378 76938-380 76938-382
6-Fluoro-4-iodo-3-pyridinol ≥97%
6-Fluoro-4-iodo-3-pyridinol

Precaución: for Research Use Only

Formula: C₅H₃FINO
MW: 238.986 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16611043
CAS Number: 1034467-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR