2-Formyl-5-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Methoxypyridine-2-carbaldehyde, 5-Methoxypicolinaldehyde

23985-1G 23985-25G 23985-5G
76938-362EA 114.58 USD
76938-362 76938-364 76938-366
2-Formyl-5-methoxypyridine ≥97%
2-Formyl-5-methoxypyridine
Formula: C₇H₇NO₂
MW: 137.1360 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08234867
CAS Number: 22187-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR