Methyl-4-fluoro-2-methoxybenzoate ≥95%

Supplier: AOBChem USA

Warning

23965-25G 23965-5G
76938-354EA 310.85 USD
76938-354 76938-356
Methyl-4-fluoro-2-methoxybenzoate ≥95%
Methyl-4-fluoro-2-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₉FO₃
Storage Temperature: Refrigerator
MDL Number: MFCD06203990
CAS Number: 204707-42-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR