6-Fluoro-2-(Methylsulfanyl)Phenylboronic Acid ≥97%

Supplier: AOBChem USA

Warning

23947-1G 23947-25G 23947-500MG 23947-5G
76938-342EA 201.58 USD
76938-342 76938-344 76938-346 76938-348
6-Fluoro-2-(Methylsulfanyl)Phenylboronic Acid ≥97%
6-Fluoro-2-(Methylsulfanyl)Phenylboronic Acid

Precaución: for Research Use Only

Formula: C₇H₈BFO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD22543677
CAS Number: 1451392-58-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR