Methyl-3-bromo-2,6-difluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 3-bromo-2,6-difluorobenzoate

23911-1G 23911-25G 23911-5G
76938-328EA 114.58 USD
76938-328 76938-330 76938-332
Methyl-3-bromo-2,6-difluorobenzoate ≥97%
Methyl-3-bromo-2,6-difluorobenzoate
Formula: C₈H₅BrF₂O₂
MW: 251.03 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09951948
CAS Number: 1378875-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR