Methyl-6-(dimethylamino)nicotinate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 6-dimethylamino-3-pyridinecarboxylate

23868-1G 23868-25G 23868-5G
76938-318EA 66.84 USD
76938-318 76938-320 76938-322
Methyl-6-(dimethylamino)nicotinate ≥97%
Methyl-6-(dimethylamino)nicotinate

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O₂
MW: 180.21 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10699718
CAS Number: 26218-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR