6-Chloro-4-iodonicotinic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Chloro-4-iodopyridine-3-carboxylic acid

23526-1G 23526-500MG 23526-5G
76938-282EA 593.05 USD
76938-282 76938-284 76938-286
6-Chloro-4-iodonicotinic acid ≥97%
6-Chloro-4-iodonicotinic acid
Formula: C₆H₃ClINO₂
MW: 283.45 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18259173
CAS Number: 1211578-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR