7-Chloro-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 7-Chloroindazole

23306-25G 23306-5G
76938-266EA 310.85 USD
76938-266 76938-268
7-Chloro-1H-indazole ≥97%
7-Chloro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₅ClN₂
MW: 152.5810 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07368214
CAS Number: 37435-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR