(7-Methyl-1H-indazol-4-yl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 7-Methyl-1H-indazole-4-boronic acid

23280-1G 23280-500MG 23280-5G
76938-254EA 846.6 USD
76938-254 76938-256 76938-258
(7-Methyl-1H-indazol-4-yl)boronic acid ≥97% research grade
(7-Methyl-1H-indazol-4-yl)boronic acid
Formula: C₈H₉BN₂O₂
MW: 175.98 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12028642
CAS Number: 1310404-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR