6-Fluoro-2-hydroxy-3-methylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

23197-1G 23197-25G 23197-500MG 23197-5G
76938-240EA 379.81 USD
76938-240 76938-242 76938-244 76938-246
6-Fluoro-2-hydroxy-3-methylphenylboronic acid pinacol ester ≥97%
6-Fluoro-2-hydroxy-3-methylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₈BFO₃
MW: 252.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22056753
CAS Number: 1451391-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR