Methyl-2-chloro-5-methylnicotinate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 2-chloro-5-methylpyridine-3-carboxylate, Methyl 2-chloro-5-methylnicotinate

23058-1G 23058-25G 23058-500MG 23058-5G
76938-216EA 162.32 USD
76938-216 76938-218 76938-220 76938-222
Methyl-2-chloro-5-methylnicotinate ≥97%
Methyl-2-chloro-5-methylnicotinate
Formula: C₈H₈ClNO₂
MW: 185.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07375372
CAS Number: 65169-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR