5-Methyl-2-isopropoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

23010-1G 23010-25G 23010-500MG 23010-5G
76938-192EA 185.66 USD
76938-192 76938-194 76938-196 76938-198
5-Methyl-2-isopropoxybenzoic acid ≥97%
5-Methyl-2-isopropoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
Storage Temperature: Refrigerator
MDL Number: MFCD14535996
CAS Number: 104216-91-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR