4-(Ethoxycarbonyl)phenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 4-(Ethoxycarbonyl)benzeneboronic acid pinacol ester, Ethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

22944-25G 22944-5G
76938-180EA 371.32 USD
76938-180 76938-182
4-(Ethoxycarbonyl)phenylboronic acid pinacol ester ≥97%
4-(Ethoxycarbonyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₁BO₄
Storage Temperature: Ambient
MDL Number: MFCD03453659
CAS Number: 195062-62-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR