7-Bromo-2,3-dihydro-1,4-benzodioxin-6-carboxaldehyde ≥97%

Supplier: AOBChem USA

Warning

22648-1G 22648-25G 22648-500MG 22648-5G
76938-150EA 213.25 USD
76938-150 76938-152 76938-154 76938-156
7-Bromo-2,3-dihydro-1,4-benzodioxin-6-carboxaldehyde ≥97%
7-Bromo-2,3-dihydro-1,4-benzodioxin-6-carboxaldehyde

Precaución: for Research Use Only

Formula: C₉H₇BrO₃
MW: 243.054 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09118354
CAS Number: 99067-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR