Methyl-2-chloro-6-(trifluoromethyl)nicotinate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 2-chloro-6-(trifluoromethyl)nicotinate

22326-1G 22326-25G 22326-500MG 22326-5G
76938-098EA 168.69 USD
76938-098 76938-100 76938-102 76938-104
Methyl-2-chloro-6-(trifluoromethyl)nicotinate ≥97%
Methyl-2-chloro-6-(trifluoromethyl)nicotinate
Formula: C₈H₅ClF₃NO₂
MW: 239.58 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14698016
CAS Number: 1073129-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR