Ethyl 5-bromo-2-ethoxynicotinate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 5-bromo-2-ethoxy-3-pyridinecarboxylate

22119-1G 22119-25G 22119-500MG 22119-5G
76938-058EA 185.66 USD
76938-058 76938-060 76938-062 76938-064
Ethyl 5-bromo-2-ethoxynicotinate ≥97%
Ethyl 5-bromo-2-ethoxynicotinate

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrNO₃
MW: 274.111 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD19105409
CAS Number: 1310416-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR