Methyl 2,4-dibromo-6-fluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

22113-1G 22113-25G 22113-500MG 22113-5G
76938-046EA 260.99 USD
76938-046 76938-048 76938-050 76938-052
Methyl 2,4-dibromo-6-fluorobenzoate ≥97%
Methyl 2,4-dibromo-6-fluorobenzoate
Formula: C₈H₅Br₂FO₂
Storage Temperature: Refrigerator
MDL Number: MFCD06204165
CAS Number: 773134-10-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR