(6-Isobutoxy-5-methylpyridin-3-yl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Isobutoxy-5-methylpyridine-3-boronic acid

21926-1G 21926-25G 21926-5G
76938-032EA 100.79 USD
76938-032 76938-034 76938-036
(6-Isobutoxy-5-methylpyridin-3-yl)boronic acid ≥97%
(6-Isobutoxy-5-methylpyridin-3-yl)boronic acid
Formula: C₁₀H₁₆BNO₃
MW: 209.05 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18087688
CAS Number: 1256355-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR