Ethyl-2,5-dichloroisonicotinate ≥95%

Supplier: AOBChem USA

Warning

21821-25G 21821-5G
76938-020EA 573.95 USD
76938-020 76938-022
Ethyl-2,5-dichloroisonicotinate ≥95%
Ethyl 2,5-dichloroisonicotinate

Precaución: for Research Use Only

Formula: C₈H₇Cl₂NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD09972252
CAS Number: 603122-76-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR