Methyl 3-fluoro-2-hydroxybenzoate ≥97%

Supplier: AOBChem USA

Warning

21721-1G 21721-25G 21721-5G
76937-978EA 93.36 USD
76937-978 76937-980 76937-982
Methyl 3-fluoro-2-hydroxybenzoate ≥97%
Methyl 3-fluoro-2-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₈H₇FO₃
Storage Temperature: Ambient
MDL Number: MFCD06203651
CAS Number: 70163-98-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR